Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Alethia™ Bordetella Pertussis External Control
GSA_VA

Catalog No. 23029009 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
18 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-029-009 18 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-029-009 Supplier Meridian Bioscience™ Supplier No. 479930
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with Alethia Pertussis DNA Amplification Assay

  • Contains positive control reagent
  • Formulation: Tris-buffered solution containing non-infectious plasmid DNA (B. pertussis target DNA inserts) with azide (0.09%) as preservative

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Type Alethia Bordetella Pertussis External Control Kit
Includes Positive control reagents
Intended Use Alethia Bordetella pertussis DNA Amplification Assay
Quantity 18 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.