Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Alethia™ Bordetella Pertussis Kit
GSA_VA

Catalog No. 23029008 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-029-008 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-029-008 Supplier Meridian Bioscience™ Supplier No. 480750
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Diagnostic system for detection of Bordetella pertussis in nasopharyngeal swab samples

Bordetella Pertussis is the pathogen responsible for the endemic respiratory disease whooping cough. Although there is a vaccine available, it is currently showing an increase of reportable infections in the US. Molecular assays are increasingly being used due to improved sensitivity and rapid turnaround time.

  • Based on loop-mediated isothermal DNA amplification (LAMP) technology
  • Result interpretation in less than 1 hour by illumipro-10 Incubator/Reader
  • High sensitivity (87.8%) and specificity (97.8%)
  • Quicker initiation of patient treatment
  • Small instrument footprint
  • Simple testing protocol, easily train all laboratory personnel

The device is not intended for point-of-care use.

Healthcare industry

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Content And Storage 2°C to 30°C
For Use With (Application) Detection of Bordetella pertussis in nasopharyngeal swab samples
Quantity 50 Tests
Sensitivity Drift 87.8%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.