Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Alethia™ CMV Assay
GSA_VA

Catalog No. 23029000 Shop All Meridian Bioscience Products
Change view
Click to view available options
Test Time:
<60 min.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Test Time
23-029-000 <60 min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-029-000 Supplier Meridian Bioscience™ Supplier No. 481325
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Meridian Bioscience™ Alethia™ CMV Assay is an in vitro diagnostic molecular test for the detection of CMV from the saliva of newborns under 21 days.

  • Report results with confidence with the First FDA cleared test for cCMV
  • Utilizes saliva, an easy to collect and preferred sample type for congenital CMV testing
  • Obtain results in less than one hour with minimal hands on time
  • High NPV provides confidence to physicians and piece of mind to parents before their baby goes home
  • The simple procedure of Alethia™ CMV provides flexibility to best fit the laboratory's workflow when compared to other testing technologies
  • Saliva is easy to collect and contains high viral load, allowing optimal detection and same day reporting for physicians and families
  • Implementing newborn testing strategies for cCMV have been increasingly recognized for their potential medical benefits and improved patient outcomes
  • Congenital CMV testing empowers physicians and families with the full understanding of their newborn's health prior to leaving the hospital

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Sample Type Neonatal Saliva Swab
For Use With (Application) Detection of Cytomegalovirus DNA
Test Time <60 min.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.