Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Alethia™ CMV External Control
GSA_VA

Catalog No. 23029001 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
10 Samples
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-029-001 10 Samples
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-029-001 Supplier Meridian Bioscience™ Supplier No. 479880
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Meridian Bioscience™ Alethia™ CMV External Control Reagents include materials for use with Alethia™ CMV DNA Amplification Assay.

  • Used as part of a routine quality control program to aid the user in detection of unexpected conditions that may lead to test errors
  • The external controls are intended for use with the Alethia™ CMV DNA Amplification Assay; the controls are not intended for use with other assays or systems.
  • Contains Alethia™ CMV Positive Control: Tris-buffered solution with plasmid containing DNA inserts (Cytomegalovirus and human mitochondrial DNA inserts) and azide (0.09%) as a preservative

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Description Alethia™ CMV Positive/Negative Controls
For Use With (Application) Alethia™ CMV DNA Amplification Assay
Quantity 10 Samples
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.