Learn More
Meridian Bioscience™ Alethia™ External Control Kit
GSA_VA
Catalog No. 23029003 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
18 Tests
Catalog No. Quantity
23-029-003 18 Tests
1 options
This item is not returnable. View return policy
Catalog No. 23-029-003 Supplier Meridian Bioscience™ Supplier No. 479920
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
For the detection of C. difficiles in stool
illumigene C. difficile External Control Reagents are supplied separately It is recommended that the reactivity of each new lot and each new shipment of illumigene C. difficile be verified on receipt and before use. External control tests should be performed thereafter in accordance with appropriate federal, state and local guidelines. The illumigene C. difficile test kit should not be used in patient testing if the external controls do not produce the correct results.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Certifications/Compliance CE Marked
Detectable Analytes C. difficile
Type Alethia External Control Kit
Includes Positive and negative control reagent
Quantity 18 Tests
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov