Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience Alethia GAS Best of Both Worlds Program
GSA_VA

Catalog No. 23029050 Shop All Meridian Bioscience Products
Change view
Click to view available options
Detectable Analytes:
Streptococci (Group A)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
23-029-050 Streptococci (Group A)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-029-050 Supplier Meridian Bioscience Supplier No. 280150P23
Add to Cart
Add to Cart
This item is not returnable. View return policy

Improves decision making for physicians by providing a convenient and timely result of Group A Strep pharyngitis.

  • The Alethia Best of Both Worlds GAS Solution includes 2 Alethia Group A Strep
  • Molecular Test Kits and 3 Immunocard STAT! Strep A Rapid test kits
  • Improves decision making for physicians by providing a convenient and timely result of Group A Strep pharyngitis
  • Test with Immunocard STAT! Strep A first and be able to treat while patient is waiting
  • Reflex Rapid Group A Strep negatives to the Alethia Group A Strep molecular to confirm results
  • Alethia GAS Turnaround Time: Less than 60 min
  • ImmunoCard STAT! Strep A Turnaround Time: Less than 5 min

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

CE Marker Yes
Clia Complexity Moderate
Detectable Analytes Streptococci (Group A)
Format Kit
Type Strep Test Kit
Disposable Single-use
For Use With (Equipment) Alethia Instrument (23-029-014)
Form Ready-to-Use (RTU)
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.