Learn More
Meridian Bioscience™ Alethia™ Group A Streptococcus Assays Control Set
GSA_VA
Catalog No. 23029005 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
18 Tests
Catalog No. Quantity
23-029-005 18 Tests
1 options
This item is not returnable. View return policy
Catalog No. 23-029-005 Supplier Meridian Bioscience™ Supplier No. 479910
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
Clinically definitive molecular amplification assay for Group A Strep
The illumigene Group A Streptococcus assay performed on the illumipro-10™ is a qualitative in vitro diagnostic test for the detection of Streptococcus pyogenes (Group A β-hemolytic Streptococcus) in throat swab specimens.
  • Simplify procedures in laboratory
  • Provide more definitive, accurate result than rapid tests or traditional cultures
  • Eliminate subjectivity associated with plate reading
  • Increase detection of positives by 53% over traditional culture in symptomatic patients
  • For use with illumipro-10™ molecular diagnostic system
  • Starter Package and Control Set also available
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Certifications/Compliance CE Marked
Detectable Analytes Streptococci (Group A)
Type Group A Streptococcus Assay
Quantity 18 Tests
Test Time 60 min.