Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Alethia™ Group A Streptococcus Assays
GSA_VA

Catalog No. 23029004 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
50 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-029-004 50 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-029-004 Supplier Meridian Bioscience™ Supplier No. 480150
Add to Cart
Add to Cart
This item is not returnable. View return policy

Clinically definitive molecular amplification assay for Group A Strep

The illumigene Group A Streptococcus assay performed on the illumipro-10™ is a qualitative in vitro diagnostic test for the detection of Streptococcus pyogenes (Group A β-hemolytic Streptococcus) in throat swab specimens.

  • Simplify procedures in laboratory
  • Provide more definitive, accurate result than rapid tests or traditional cultures
  • Eliminate subjectivity associated with plate reading
  • Increase detection of positives by 53% over traditional culture in symptomatic patients
  • For use with illumipro-10™ molecular diagnostic system
  • Starter Package and Control Set also available

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance CE Marked
CPT Code 87651
Detectable Analytes S. pyogenes
Sample Type Throat Swab
Type Group A Streptococcus Assay
Quantity 50 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.