Learn More
Meridian Bioscience™ Alethia™ Group B Streptococcus Test Controls
GSA_VA
Catalog No. 23029007 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
18 Tests
Catalog No. Quantity
23-029-007 18 Tests
1 options
This item is not returnable. View return policy
Catalog No. 23-029-007 Supplier Meridian Bioscience™ Supplier No. 479900
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
Clinically definitive molecular amplification assay for Group B Strep
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Content And Storage 2°C to 8°C
Type Isothermal Nucleic Acid Amplification Assay
Quantity 18 Tests