Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Alethia™ HSV 1 and 2 DNA Amplification Assay
GSA_VA

Catalog No. 23029010 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
25 Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-029-010 25 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-029-010 Supplier Meridian Bioscience™ Supplier No. 480650
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

25 Tests each for HSV-1 and HSV-2.

Qualitative in vitro diagnostic test for direct detection and differentiation of herpes simplex virus type 1 and type 2 in cutaneous and mucocutaneous lesion specimens.

illumigene HSV 1&2 utilizes loop-mediated isothermal DNA amplification (LAMP) to detect HSV-1 and HSV-2. Results are available in less than one hour to reduce turn around time, eliminate send-out costs and aid in same day diagnosis, treatment and counseling of patients.
  • Results in less than one hour
  • High sensitivity and specificity improves accuracy of diagnosis
  • Testing requires <2 minutes of hands-on time
  • Expanded molecular menu
  • Reduce cost and increase profitability
  • No capital expense or service contract required
  • Moderately complex with minimal hands-on time
  • Small footprint, easily adaptable to any laboratory
Specifications:
  • HSV-1 Sensitivity: Cutaneous: 94.1%, Mucocutaneous: 95.0%
  • HSV-2 Sensitivity: Cutaneous: 100%, Mucocutaneous: 98.6%
  • HSV-1 Specificity: Cutaneous: 97.2%, Mucocutaneous: 94.9%
  • HSV-2 Specificity: Cutaneous: 95.1%, Mucocutaneous: 95.6%
  • Acceptable Sample Types: Cutaneous: skin and genital (penis); Mucocutaneous: anorectal and genital (vaginal/cervical, nasal, ocular, oral, urethral)
  • Assay Format: Isothermal DNA amplification

Certifications

CE marked

Storage: Store kits at 2° to 30°C and external controls at 2° to 8°C.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance CE Marked
Content And Storage Store kits at 2°C to 30°C and external controls at 2°C to 8°C.
Clia Complexity Moderate
Detectable Analytes HSV 1/2
Sample Type Mucocutaneous and Cutaneous
Sensitivity HSV-1: Cutaneous: 94.1%, Mucocutaneous: 95.0%; HSV-2: Cutaneous: 100%, Mucocutaneous: 98.6%
Type HSV 1& DNA Amplification Assay
For Use With (Application) Detects and differentiates HSV-1 and HSV-2
Includes 25 Tests each for HSV-1 and HSV-2
Quantity 25 Each
Specificity HSV-1: Cutaneous: 97.2%, Mucocutaneous: 94.9%; HSV-2: Cutaneous: 95.1%, Mucocutaneous: 95.6%
Test Time <1 hr.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.