Learn More
Medchemexpress LLC Aleuria Aurantia Lectin (AAL) | 2 MG
SDP
Catalog No. 5000426526
Encompass_Preferred
Aleuria Aurantia Lectin (AAL) is a plant lectin and a biological material or organic compound. It serves as a probe to specifically bind biomolecules, such as polysaccharides and peptides. This product is intended for research use only.
  • Used as a probe to specifically bind biomolecules
  • Binds to polysaccharides and peptides
  • Suitable for life science research
Catalog No. 50-004-26526 Supplier Medchemexpress LLC Supplier No. HYNP0632MG
Aleuria Aurantia Lectin (AAL) is a plant lectin and a biological material or organic compound. It serves as a probe to specifically bind biomolecules, such as polysaccharides and peptides. This product is intended for research use only.
  • Used as a probe to specifically bind biomolecules
  • Binds to polysaccharides and peptides
  • Suitable for life science research