Learn More
Medchemexpress LLC Aleuria aurantia lectin (biotinylated) | 1 MG
SDP
Catalog No. 5000425338
Encompass_Preferred
Aleuria Aurantia Lectin (AAL) Biotinylated is a plant lectin that serves as a probe to specifically bind biomolecules such as polysaccharides and peptides. It is a biological material or organic compound intended for use in life science research.
  • Functions as a plant lectin
  • Serves as a probe to specifically bind biomolecules
  • Capable of binding polysaccharides and peptides
  • Applicable as a biological material or organic compound
  • Intended for use in life science research
Catalog No. 50-004-25338 Supplier Medchemexpress LLC Supplier No. HYNP01731MG
Aleuria Aurantia Lectin (AAL) Biotinylated is a plant lectin that serves as a probe to specifically bind biomolecules such as polysaccharides and peptides. It is a biological material or organic compound intended for use in life science research.
  • Functions as a plant lectin
  • Serves as a probe to specifically bind biomolecules
  • Capable of binding polysaccharides and peptides
  • Applicable as a biological material or organic compound
  • Intended for use in life science research