Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aleuritic Acid 98.0+%, TCI America™
SDP

Catalog No. A12935G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A12935G Supplier TCI America Supplier No. A12935G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 533-87-9
Molecular Formula C16H32O5
Molecular Weight (g/mol) 304.427
MDL Number MFCD00135596
InChI Key MEHUJCGAYMDLEL-LSDHHAIUSA-N
Synonym aleuretic acid, 8,9,15-trihydroxypentadecane-1-carboxylic acid, 9,10,16-trihydroxypalmitic acid, 10r,9s-9,10,16-trihydroxyhexadecanoic acid, 9s,10r-9,10,16-trihydroxyhexadecanoic acid, unii-442lq2k03a component
PubChem CID 7269316
IUPAC Name (9S,10R)-9,10,16-trihydroxyhexadecanoic acid
SMILES C(CCCC(C(CCCCCCO)O)O)CCCC(=O)O

Specifications

Melting Point 98°C
Color White-Yellow
Quantity 5 g
Formula Weight 304.43
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Aleuritic Acid
RTECSNumber ML9850000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.