Learn More
Biolegend Alexa Fluor 488 anti-human CD63
Supplier:  Biolegend 353037
Encompass_Preferred
Alexa Fluor 488 anti-human CD63 [H5C6]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. NC1612826