Learn More
Biolegend Alexa Fluor 488 anti-mouse Ly-6G
Supplier:  Biolegend 127625
Encompass_Preferred
Alexa Fluor 488 anti-mouse Ly-6G [1A8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC-F; Size: 25 ug
Catalog No. NC1730305