Learn More
Biolegend Alexa Fluor 647 anti-mouse CLEC4F
Supplier:  Biolegend 156804
Encompass_Preferred
Alexa Fluor 647 anti-mouse CLEC4F [3E3F9]; Isotype: Mouse IgG1, κ; Reactivity: Mouse, Rat; Apps: IHC-F; Size: 100 ug
Catalog No. NC1785654