Learn More
Jackson Immuno Research Labs ALEXA FLU 647 AFFINPU GO 1.5MG
SDP
Supplier:  Jackson Immuno Research Labs 102605003
Encompass_Preferred
Target: CatHost: GoatAntibody Format: Whole IgG 1.5ml33Specificity: IgG (H+L)Conjugate: Alexa Fluor® 647Product Category: Whole IgG Affinity-Purified AntibodiesClonality: PolyclonalRRID: AB_2337376
Catalog No. NC2606353