Learn More
Jackson Immuno Research Labs Alexa Fluor 488 AffiniPure Donkey Anti-Human IgG, Fcγ fragment specific | 1.0mg
SDP
Catalog No. NC1557994
Encompass_Preferred
Target: Human | Host: Donkey | Antibody Format: Whole IgG | Specificity: IgG, Fcγ fragment specific | Minimal Cross Reactivity: Bovine, Horse, Mouse Serum Proteins | Conjugate: Alexa Fluor 488
Catalog No. NC1557994 Supplier Jackson Immuno Research Labs Supplier No. 709545098
Target: Human | Host: Donkey | Antibody Format: Whole IgG | Specificity: IgG, Fcγ fragment specific | Minimal Cross Reactivity: Bovine, Horse, Mouse Serum Proteins | Conjugate: Alexa Fluor 488