Learn More
Jackson Immuno Research Labs Alexa Fluor 488 AffiniPure Goat Anti-Human IgG, F(ab')₂ fragment specific, 1mg
SDP
Supplier:  Jackson Immuno Research Labs 109545097
Encompass_Preferred
Alexa Fluor 488-AffiniPure Goat Anti-Human IgG, F(ab')2 Fragment Specific (min X Bov,Hrs,Ms Sr Prot)
Catalog No. NC0555641