Learn More
Biolegend Alexa Fluor 488 anti-DYKDDDDK Tag Antibody, 25 ug
Supplier:  Biolegend 637317
Encompass_Preferred
The DYKDDDDK tag, commonly referred to as Sigma's FLAG Tag, is often used as a protein modification in order to simplify the labeling and detection of proteins.This unique amino acid sequence allows for specific antibody detection in western blotting, immunoprecipitation, and immunostaining techniques.Due to the short sequence, this modification is not likely to affect the structure or function of the modified proteins.
Catalog No. NC1908756