Learn More
Biolegend Alexa Fluor 488 anti-His Tag
Supplier:  Biolegend 652509
Encompass_Preferred
Alexa Fluor 488 anti-His Tag [J099B12]; Isotype: Mouse IgG1, κ; Reactivity: All species (6x His tag epitope); Apps: ICC; Size: 25 ug
Catalog No. NC1574884