Learn More
Biolegend Alexa Fluor 488 anti-human FOXP3
Supplier:  Biolegend 320212
Encompass_Preferred
Alexa Fluor 488 anti-human FOXP3 [259D]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus, Rhesus, Baboon, Chimpanzee; Apps: ICFC; Size: 100 tests
Catalog No. NC0904384