Learn More
Jackson Immuno Research Labs Alexa Fluor 488 AffiniPure Donkey Anti-Human IgG (H+L), .5mg
SDP
Catalog No. NC0179262
Encompass_Preferred
Alexa Fluor 488-AffiniPure Donkey Anti-Human IgG (H+L) (min X Bov,Ck,Gt,GP,Sy Hms,Hrs,Ms,Rb,Rat,Shp Sr Prot)
Catalog No. NC0179262 Supplier Jackson Immuno Research Labs Supplier No. 709545149
Alexa Fluor 488-AffiniPure Donkey Anti-Human IgG (H+L) (min X Bov,Ck,Gt,GP,Sy Hms,Hrs,Ms,Rb,Rat,Shp Sr Prot)