Learn More
Biolegend Alexa Fluor 488 anti-human CD326 (Ep-CAM)
Supplier:  Biolegend 324210
Encompass_Preferred
Alexa Fluor 488 anti-human CD326 (Ep-CAM) [9C4]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1344332