Learn More
Abcam Ale x a Fluor 488 Conjugation Kit (Fast) - Lightning-Link, 1MG
Catalog No. NC2019756
Encompass_Preferred
Alexa Fluor 488 protein labeling/conjugation kit, Lightning-Link requires < 20 minutes with 30 seconds hands-on time. - Cited in over 30 publications - 100% antibody recovery - 15-minute incubation time Custom-size conjugation kits up to 100 mg are available on demand.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2019756 Supplier Abcam Supplier No. AB2365531MG
Alexa Fluor 488 protein labeling/conjugation kit, Lightning-Link requires < 20 minutes with 30 seconds hands-on time. - Cited in over 30 publications - 100% antibody recovery - 15-minute incubation time Custom-size conjugation kits up to 100 mg are available on demand.
The product is subject to the following: Abcam Restricted Use Statement