Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Alexa Fluor 488-Labeled Monoclonal Anti-G4S Linker Antibody, Rabbit IgG (Clone 016) - 25 tests

Catalog No. NC3907853
Encompass_Preferred

Alexa Fluor 488-labeled monoclonal anti-G4S linker antibody (rabbit IgG, clone 016) is optimized for detection of G4S and G3S linkers in recombinant proteins and CAR scFv constructs by flow cytometry. The reagent is supplied lyophilized for convenient storage and reconstitution.

  • Recognizes (G3S)n and (G4S)n linkers with a minimum of two repeats.
  • Label: Alexa Fluor 647; excitation 640 nm, emission 672 nm.
  • Host species: rabbit; isotype: IgG, kappa.
  • Size: 25 tests; recommended dilution 1:50.
  • Formulation: lyophilized from 0.22 μm filtered PBS with trehalose.
  • Preservative: 0.03% Proclin 300.
  • Storage: lyophilized -20°C to -70°C for 24 months; -70°C for 12 months after reconstitution.
  • Intended use: flow cytometry evaluation of cell-surface CARs or proteins containing G4S linkers.

Catalog No. NC3907853 Supplier ACROBiosystems Supplier No. G4SAFYP125TESTS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alexa Fluor 488-labeled monoclonal anti-G4S linker antibody (rabbit IgG, clone 016) is optimized for detection of G4S and G3S linkers in recombinant proteins and CAR scFv constructs by flow cytometry. The reagent is supplied lyophilized for convenient storage and reconstitution.

  • Recognizes (G3S)n and (G4S)n linkers with a minimum of two repeats.
  • Label: Alexa Fluor 647; excitation 640 nm, emission 672 nm.
  • Host species: rabbit; isotype: IgG, kappa.
  • Size: 25 tests; recommended dilution 1:50.
  • Formulation: lyophilized from 0.22 μm filtered PBS with trehalose.
  • Preservative: 0.03% Proclin 300.
  • Storage: lyophilized -20°C to -70°C for 24 months; -70°C for 12 months after reconstitution.
  • Intended use: flow cytometry evaluation of cell-surface CARs or proteins containing G4S linkers.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.