Learn More
ACROBiosystems Alexa Fluor™ 488-Labeled Human Alpha-synuclein Protein, Tag Free, 200ug
Catalog No. NC3735145
Encompass_Preferred
Alexa Fluor 488-Labeled Human Alpha-synuclein Protein, Tag Free(ALN-HA113) is expressed from E. coli cells. It contains AA Met 1 - Ala 140 (Accession # P37840-1).Predicted N-terminus: Met 1
Catalog No. NC3735145 Supplier ACROBiosystems Supplier No. ALNHA113200UG
Alexa Fluor 488-Labeled Human Alpha-synuclein Protein, Tag Free(ALN-HA113) is expressed from E. coli cells. It contains AA Met 1 - Ala 140 (Accession # P37840-1).Predicted N-terminus: Met 1