Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Alexa Fluor™ 555 Cadaverine

Catalog No. A30677
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30677 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A30677 Supplier Invitrogen™ Supplier No. A30677
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Alexa Fluor™ 555 Cadaverine is useful as a polar tracer and as a reactive dye for labeling proteins via a carboxylic acid moiety.

Alexa Fluor™ 555 Cadaverine is useful as a polar tracer and as a reactive dye for labeling proteins via a carboxylic acid moiety. Alexa Fluor™ 555 is a bright, orange dye. Used for stable signal generation in imaging and flow cytometry, Alexa Fluor™ 555 dye is water soluble and pH-insensitive from pH 4 to pH 10. In addition to reactive dye formulations, we offer Alexa Fluor™ 555 dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection.

Detailed information about this AlexaFluor™ cadaverine:
• Fluorophore label : Alexa Fluor™ 555 dye
• Reactive group: cadaverine
• Reactivity: carboxylic acids, aldehydes, and ketones (and glutamine residues through an enzyme-catalyzed transamidation reaction)
• Ex/Em of the conjugate: 555/572 nm
• Extinction coefficient: 155,000 cm-1M-1
• Spectrally similar dyes: TRITC
• Molecular weight: ∼950

Cell Tracking and Tracing Applications
Alexa Fluor™ cadaverines make excellent fluorescent polar tracers because they are bright, small, and water soluble. Since they contain an aldehyde-fixable functional group, they can be fixed in cells by treatment with formaldehyde or glutaraldehyde. They are easily loaded into cells by microinjection, infusion from patch pipette, or uptake induced by our Influx™ Pinocytic Cell-Loading Reagent (Cat. No. I14402).

Protein Labeling Applications
Alexa Fluor™ cadaverines can be used as reactive molecules for adding a fluorescent label to carboxylic acids using a coupling agent such as a carbodiimide; they do not spontaneously react with carboxylic acids in solution. They do, however, react spontaneously with the common amine-reactive functional groups, including succinimidyl esters and isothiocyanates. The amine-containing Alexa Fluor™ cadaverines can also be used to label glutamine residues in some proteins and peptides via an enzyme-catalyzed transamidation reaction.

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We’ll Make a Custom Conjugate for You
If you can’t find what you’re looking for in our online catalog, we’ll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

Related Products
DMSO (dimethylsulfoxide) (D12345)
Antibody Conjugate Purification Kit for 0.5-1 mg (A33086)
Antibody Conjugate Purification Kit for 20-50 μg (A33087)
Antibody Conjugate Purification kit for 50-100 μg (A33088)

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Product Type Cadaverine
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Chemical Reactivity Carboxylic Acid, Ketone, Aldehyde
Shipping Condition Room Temperature
Reactive Moiety Amine, Cadaverine
Label or Dye Alexa Fluor™ 555
Label Type Alexa Fluor
Product Line Alexa Fluor
Emission 572 nm
Excitation 555 nm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.