Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Alexa Fluor™ 594 Alkyne (Alexa Fluor™ 594 Carboxamido-(5-(and 6-)Propargyl), Bis(Triethylammonium Salt)), mixed isomers

Catalog No. 501148693
Encompass_Preferred
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-114-8693 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-114-8693 Supplier Invitrogen™ Supplier No. A10275
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Alexa Fluor™ 594 Alkyne (Alexa Fluor™ 594 Carboxamido-(5-(and 6-)Propargyl), Bis(Triethylammonium Salt)), mixed isomers

The red-fluorescent Alexa Fluor™ 594 alkyne can be reacted with azides via a copper-catalyzed click reaction. Click chemistry describes a class of chemical reactions that use bio-orthogonal or biologically unique moities to label and detect a molecule of interest using a two-step procedure. The two-step reaction procedure involves a copper-catalyzed triazole formation of an azide and an alkyne. Click reactions have several characteristics: the reaction between the detection moieties is efficient; no extreme temperatures or solvents are required; the reaction product is stable; the components of the reaction are bioinert; and perhaps most importantly, no side reactions occur – the label and detection tags react selectively and specifically with one another. Unlike traditional chemical reactions utilizing succinimidyl esters or maleimides that target amines and sulfhydryls – functional groups that are not unique – click chemistry-labeled molecules can be applied to complex biological samples and be detected with unprecedented sensitivity due to extremely low background.
TRUSTED_SUSTAINABILITY

Specifications

Format Solid
Quantity 0.5 mg
Product Type Alkyne
Molecular Weight (g/mol) 962.63 Da
Solubility DMSO (Dimethylsulfoxide)
Content And Storage Store at ≤-20°C, desiccate and protect from light.
Chemical Reactivity Azide
Detection Method Fluorescence
Reactive Group Alkyne
Shipping Condition Room Temperature
Reactive Moiety Alkyne
Label or Dye Alexa Fluor™ 594
Label Type Alexa Fluor
Color Red
Product Line Alexa Fluor, Molecular Probes
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.