Learn More
Biolegend Alexa Fluor 594 anti-c-Myc
Supplier:  Biolegend 626805
Encompass_Preferred
Alexa Fluor 594 anti-c-Myc [9E10]; Isotype: Mouse IgG1; Reactivity: Endogenous Human c-Myc; fusion protein with c-Myc tag (EQKLISEEDL, around 1.2kD) in all species; Apps: ICC; Size: 25 ug
Catalog No. NC1676422