Learn More
Biolegend Alexa Fluor 594 anti-human CD20
Supplier:  Biolegend 302354
Encompass_Preferred
Alexa Fluor 594 anti-human CD20 [2H7]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Baboon, Capuchin Monkey, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus, Squirrel Monkey; Apps: ICC, IHC-F; Size: 100 ug
Catalog No. NC1417740