Learn More
Biolegend Alexa Fluor 594 anti-Nestin
Supplier:  Biolegend 655106
Encompass_Preferred
Alexa Fluor 594 anti-Nestin [Rat-401]; Isotype: Mouse IgG1, κ; Reactivity: Mouse, Rat; Apps: IHC-P; Size: 100 ug
Catalog No. 50-208-7075