Learn More
Abcam Ale x a Fluor 594 Conjugation Kit (Fast) - Lightning-Link, 3 x 10UG
Catalog No. NC2085369
Encompass_Preferred
Alexa Fluor 594 conjugation / labeling in < 20 mins with 30 secs hands-on time using Alexa Fluor 594 Conjugation Kit (Fast) - Lightning-Link ab269822. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
This item is not returnable. View return policy
Catalog No. NC2085369 Supplier Abcam Supplier No. AB2698223X10UG
Alexa Fluor 594 conjugation / labeling in < 20 mins with 30 secs hands-on time using Alexa Fluor 594 Conjugation Kit (Fast) - Lightning-Link ab269822. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement