Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Alexa Fluor™ 594 Hydrazide, for microinjection

Catalog No. A10442
Encompass_Preferred
Change view
Click to view available options
Quantity:
125 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10442 125 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A10442 Supplier Invitrogen™ Supplier No. A10442
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Alexa Fluor™ 594 Hydrazide is useful as a cell tracer and as a reactive dye for labeling aldehydes or ketones in polysaccharides or glycoproteins.

Alexa Fluor™ 594 Hydrazide is useful as a cell tracer and as a reactive dye for labeling aldehydes or ketones in polysaccharides or glycoproteins. This version is formatted as a ready-to-use solution that is dissolved in a 200 mM KCl solution and filter sterilized.

Alexa Fluor™ 594 is a bright, red fluorescent dye. Used for stable signal generation in imaging and flow cytometry, Alexa Fluor™ 594 dye is water soluble and pH-insensitive from pH 4 to pH 10. In addition to reactive dye formulations, we offer Alexa Fluor™ 594 dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection.

Detailed information about this AlexaFluor™ hydrazide:
• Fluorophore label : Alexa Fluor™ 594 dye
• Reactive group: hydrazide
• Reactivity: Aldehydes or keytones in polysaccharides or glycoproteins
• Ex/Em of the conjugate: 588/613 nm
• Extinction coefficient: 97,000 cm-1M-1
• Spectrally similar dyes: Texas Red
• Molecular weight: 758.79

Cell Tracking and Tracing Applications
Alexa Fluor™ hydrazides and hydroxlamines are useful as low molecular weight, membrane-impermeant, aldehyde-fixable cell tracers, exhibiting brighter fluorescence and greater photostability than cell tracers derived from other spectrally similar fluorophores. They are easily loaded into cells by microinjection, infusion from patch pipette, or uptake induced by our Influx™ Pinocytic Cell-Loading Reagent (Cat. No. I14402). Learn more about cell tracking and tracing.

Glycoprotein and Polysaccharide Labeling Applications
The Alexa Fluor™ hydrazides and hydroxlamines are reactive molecules that can be used to add a fluorescent label to biomolecules containing aldehydes or ketones. Aldehydes and ketones can be introduced into polysaccharides and glycoproteins by periodate-mediated oxidation of vicinal diols. Galactose oxidase can also be used to oxidize terminal galactose residues of glycoproteins to aldehydes.

Hydrazide vs Hydroxylamine
Hydrazine derivatives react with ketones and aldehydes to yield relatively stable hydrazones. Hydroxylamine derivatives (aminooxy compounds) react with aldehydes and ketones to yield oximes. Oximes are superior to hydrazones with respect to hydrolytic stability. Both hydrazones and oximes can be reduced with sodium borohydride (NaBH4) to further increase the stability of the linkage.

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We’ll Make a Custom Conjugate for You
If you can’t find what you’re looking for in our online catalog, we’ll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

Related Products
DMSO (dimethylsulfoxide) (D12345)
Antibody Conjugate Purification Kit for 0.5-1 mg (A33086)
Antibody Conjugate Purification Kit for 20-50 μg (A33087)
Antibody Conjugate Purification kit for 50-100 μg (A33088)

TRUSTED_SUSTAINABILITY

Specifications

Quantity 125 μL
Product Type Hydrazide
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Carboxylic Acid, Ketone, Aldehyde
Shipping Condition Room Temperature
Reactive Moiety Amine, Hydrazide
Label or Dye Alexa Fluor™ 594
Label Type Alexa Fluor
Product Line Alexa Fluor
Emission 613 nm
Excitation 588 nm
Show More Show Less
What dye can I use that is non-reactive and can show an injection site?

The non-reactive Alexa Fluor and Alexa Fluor hydrazide derivatives may be used for injection site visualization. Other options include the fluorescent polystyrene microspheres, FluoSpheres, and dye-conjugated dextrans. The hydrazide derivatives and 'fixable' dextrans are retained by cross-linking using an aldehyde-based fixative.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.