Learn More
Biolegend Alexa Fluor 647 anti-human CD70
Supplier:  Biolegend 355116
Encompass_Preferred
Alexa Fluor 647 anti-human CD70 [113-16]; Isotype: Mouse IgG1; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1918653