Learn More
Biolegend Alexa Fluor 647 anti-human CD269 (BCMA)
Supplier:  Biolegend 357518
Encompass_Preferred
Alexa Fluor 647 anti-human CD269 (BCMA) [19F2]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1335712