Learn More
Biolegend Alexa Fluor 647 anti-human CD307c/FcRL3
Supplier:  Biolegend 374408
Encompass_Preferred
Alexa Fluor 647 anti-human CD307c/FcRL3 [H5/FcRL3]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1694269