Learn More
Biolegend Alexa Fluor 647 anti-rat IgG2b
Supplier:  Biolegend 408209
Encompass_Preferred
Alexa Fluor 647 anti-rat IgG2b [MRG2b-85]; Isotype: Mouse IgG1, κ; Reactivity: Rat; Apps: IHC-F; Size: 100 ug
Catalog No. NC1565826