Learn More
Biolegend Alexa Fluor 647 anti-Vimentin
Supplier:  Biolegend 677807
Encompass_Preferred
Alexa Fluor 647 anti-Vimentin [O91D3]; Isotype: Mouse IgG2a; Reactivity: Human; Apps: ICC, IHC-P; Size: 25 ug
Catalog No. NC1557099