Learn More
Jackson Immuno Research Labs ALEXA FLUOR 647 AP GOAT 1.0MG
SDP
Catalog No. NC1942087
Encompass_Preferred
Alexa Fluor 647 AffiniPure Goat Anti-Human IgG, Fcγ fragment specific. arget: HumanHost: GoatAntibody Format: Whole IgGSpecificity: IgG, Fcγ fragment specificMinimal Cross Reactivity: Bovine, Mouse Serum ProteinsConjugate: Alexa Fluor® 647Product Category: Whole IgG Affinity-Purified AntibodiesClonality: Polyclonal
Catalog No. NC1942087 Supplier Jackson Immuno Research Labs Supplier No. 109605190
Alexa Fluor 647 AffiniPure Goat Anti-Human IgG, Fcγ fragment specific. arget: HumanHost: GoatAntibody Format: Whole IgGSpecificity: IgG, Fcγ fragment specificMinimal Cross Reactivity: Bovine, Mouse Serum ProteinsConjugate: Alexa Fluor® 647Product Category: Whole IgG Affinity-Purified AntibodiesClonality: Polyclonal