Learn More
Abcam Alexa Fluor 647 Conjugation Kit (Fast), Pack Size 3 x 10 μg, Conjugation Target Primary amine groups, Conjugation Time Under 20 mins, Hands-on Time 30 secs
Catalog No. NC3363756
Encompass_Preferred
The Abcam Alexa Fluor 647 Conjugation Kit (Fast) provides an efficient method for labeling antibodies and proteins with Alexa Fluor 647. This kit features a quick 30-second hands-on procedure to produce ready-to-use conjugates in less than 20 minutes, without the need for further purification. Designed for primary amine groups, it ensures optimal antibody recovery and is suitable for various applications like Western blotting, ELISA, and immunohistochemistry.
  • Rapid conjugation in under 20 minutes
  • 100% antibody recovery, minimizing sample loss
  • Versatile for IgG antibodies and other proteins containing primary amine groups
  • Conjugates ready for immediate use in downstream applications
  • Scalable protocol for antibody amounts from 10 μg to 100 mg
  • Cited in over 40 scientific publications for validated performance
Catalog No. NC3363756 Supplier Abcam Supplier No. AB2698233X10UG
The Abcam Alexa Fluor 647 Conjugation Kit (Fast) provides an efficient method for labeling antibodies and proteins with Alexa Fluor 647. This kit features a quick 30-second hands-on procedure to produce ready-to-use conjugates in less than 20 minutes, without the need for further purification. Designed for primary amine groups, it ensures optimal antibody recovery and is suitable for various applications like Western blotting, ELISA, and immunohistochemistry.
  • Rapid conjugation in under 20 minutes
  • 100% antibody recovery, minimizing sample loss
  • Versatile for IgG antibodies and other proteins containing primary amine groups
  • Conjugates ready for immediate use in downstream applications
  • Scalable protocol for antibody amounts from 10 μg to 100 mg
  • Cited in over 40 scientific publications for validated performance