Learn More
Abcam Ale x a Fluor 647 Conjugation Kit (Fast) - Lightning-Link, 3 x 100UG
Catalog No. NC1882915
Encompass_Preferred
Alexa Fluor 647 antibody labeling/conjugation kit, Lightning-Link requires < 20 minutes with 30 seconds of hands-on time. 100% antibody recovery. - Cited in over 40 publications - 15-minute incubation time - Alexa Fluor 647 labeled antibody is ready for use in applications like WB, ELISA, and IHC, with no need for further purification - Compatible with most standard antibody formulations - Scalable; use the same conjugation method from 10µg-100mg of antibody - Use Lightning-Link conjugation with over 45 labels, including Alexa Fluor, DyLight, tandem dyes, enzymes, oligonucleotides, and gold nanoparticles
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1882915 Supplier Abcam Supplier No. AB2698233X100UG
Alexa Fluor 647 antibody labeling/conjugation kit, Lightning-Link requires < 20 minutes with 30 seconds of hands-on time. 100% antibody recovery. - Cited in over 40 publications - 15-minute incubation time - Alexa Fluor 647 labeled antibody is ready for use in applications like WB, ELISA, and IHC, with no need for further purification - Compatible with most standard antibody formulations - Scalable; use the same conjugation method from 10µg-100mg of antibody - Use Lightning-Link conjugation with over 45 labels, including Alexa Fluor, DyLight, tandem dyes, enzymes, oligonucleotides, and gold nanoparticles
The product is subject to the following: Abcam Restricted Use Statement