Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Alexa Fluor™ 647-DYKDDDDK Peptide

Catalog No. PV6036
Encompass_Preferred
Change view
Click to view available options
Quantity:
0.4 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV6036 0.4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV6036 Supplier Invitrogen™ Supplier No. PV6036
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alexa Fluor™ 647-DYKDDDDK Peptide

A control reagent for use with the Eu-anti-DYKDDDDK Antibody (PV6026 and PV6027).

TRUSTED_SUSTAINABILITY

Specifications

Product Line Alexa Fluor
Color Far-Red
Content And Storage 400 nM Alexa Fluor 647-DYKDDDDK Peptide in 10 mM HEPES, 137 mM NaCl, 2.7 mM KCl, pH 7.5 (HEPES buffered saline). Store at -20°C and protect from light.
Format Frozen
Label or Dye Alexa Fluor™ 647
Label Type Alexa Fluor Dyes
Quantity 0.4 mL
Research Category Drug Discovery, Immunology
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.