Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Alexa Fluor™ 647 Goat Anti-Rabbit SFX Kit, highly cross-adsorbed

Catalog No. A31634
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A31634 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A31634 Supplier Invitrogen™ Supplier No. A31634
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Alexa Fluor™ 647 Goat Anti-Rabbit SFX Kit, highly cross-adsorbed

Alexa Fluor™ SFX Kits contain Image-iT™ FX signal enhancer (Cat. no. I36933) plus one of sixteen different Alexa Fluor™ dye-labeled secondary antibodies. These kits provide 400μg (0.2 mL of 2 mg/mL) of either goat anti-mouse IgG or goat anti-rabbit IgG antibody (as a standard or highly cross-adsorbed preparation) conjugated to Alexa Fluor™ 488, Alexa Fluor™ 555, Alexa Fluor™ 594, or Alexa Fluor™ 647 dye, four of our most commonly used Alexa Fluor™ dyes. The Alexa Fluor™ 647 goat anti-rabbit IgG, highly cross-adsorbed, included with this kit is also available in a 1000μg unit size (Cat. no. A21245).
TRUSTED_SUSTAINABILITY

Specifications

Conjugate Alexa Fluor 647
Description Alexa Fluor™ 647 Goat Anti-Rabbit SFX Kit, 1kit, Goat Host, Rabbit Species Reactivity, Alexa Fluor™ 647 Conjugate, Isotype, 2 to 8°C Storage Temperature, Room Temperature Shipping Condition
Host Species Goat
Quantity 1 Kit
Regulatory Status RUO
Whole Molecule Whole Antibody
Primary or Secondary Secondary
Packaging 1 Each
Target Species Rabbit
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Reactivity Rabbit
Shipping Condition Room Temperature
Product Line Alexa Fluor, Molecular Probes
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.