Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Monoclonal Anti-G4S Linker Antibody, Alexa Fluor™ 647, Rabbit IgG, 25tests

Catalog No. NC3907851
Encompass_Preferred

This Alexa Fluor™ 647-labeled recombinant monoclonal antibody specifically recognizes proteins or scFv fragments containing (G3S)n or (G4S)n linkers, where n ≥ 2. It is expressed in human 293 cells (HEK293) to ensure high batch consistency and reliable long-term supply. The antibody is supplied as a lyophilized powder formulated in PBS with 0.03% ProClin 300 and trehalose as a protectant.

  • Host species: Rabbit
  • Isotype: Rabbit IgG, Kappa
  • Clone: 016
  • Conjugate: AF647
  • Excitation wavelength: 640 nm
  • Emission wavelength: 672 nm
  • Recommended application: Flow cytometry (1:50 dilution)
  • Reactivity: All

Catalog No. NC3907851 Supplier ACROBiosystems Supplier No. G4SABFYP125TESTS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Alexa Fluor™ 647-labeled recombinant monoclonal antibody specifically recognizes proteins or scFv fragments containing (G3S)n or (G4S)n linkers, where n ≥ 2. It is expressed in human 293 cells (HEK293) to ensure high batch consistency and reliable long-term supply. The antibody is supplied as a lyophilized powder formulated in PBS with 0.03% ProClin 300 and trehalose as a protectant.

  • Host species: Rabbit
  • Isotype: Rabbit IgG, Kappa
  • Clone: 016
  • Conjugate: AF647
  • Excitation wavelength: 640 nm
  • Emission wavelength: 672 nm
  • Recommended application: Flow cytometry (1:50 dilution)
  • Reactivity: All

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.