Learn More
Medchemexpress LLC Alexa fluor 647 NHS ester | 1620475-28-6 | 95.0% | 942.06 | 5 MG
SDP
Catalog No. 5000429227
Encompass_Preferred
Alexa fluor 647 NHS ester can be used to label Alexa fluor 647 to the primary amines (R-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules. It is for research use only and not sold to patients.
  • Used to label primary amines
  • Suitable for proteins, amine-modified oligonucleotides, and other amine-containing molecules
  • For research use only
Catalog No. 50-004-29227 Supplier Medchemexpress LLC Supplier No. HYD20965MG
Alexa fluor 647 NHS ester can be used to label Alexa fluor 647 to the primary amines (R-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules. It is for research use only and not sold to patients.
  • Used to label primary amines
  • Suitable for proteins, amine-modified oligonucleotides, and other amine-containing molecules
  • For research use only