Learn More
Medchemexpress LLC Alexa fluor 647 NHS ester | 1620475-28-6 | 95.0% | 942.06 g/mol | C39H47N3O16S4 | 10 MG
SDP
Catalog No. 5000297834
Encompass_Preferred
Alexa Fluor 647 NHS ester is a far-red, amine-reactive fluorescent dye (succinimidyl ester) used to covalently label primary amines on proteins, peptides, and amine-modified oligonucleotides for fluorescence imaging and flow cytometry.
  • Far-red, bright, and photostable fluorescent dye suitable for imaging and flow cytometry.
  • Amine-reactive NHS (succinimidyl) ester enables covalent labeling of primary amines.
  • Compatible with proteins, peptides, and amine-modified oligonucleotides.
  • High purity (95.0%) for reliable conjugation performance.
  • Supplied as a solid; store sealed at -20°C, protected from moisture and light.
  • COA, SDS, and handling documents available to support safe use and quality verification.
Catalog No. 50-002-97834 Supplier Medchemexpress LLC Supplier No. HYD209610MG
Alexa Fluor 647 NHS ester is a far-red, amine-reactive fluorescent dye (succinimidyl ester) used to covalently label primary amines on proteins, peptides, and amine-modified oligonucleotides for fluorescence imaging and flow cytometry.
  • Far-red, bright, and photostable fluorescent dye suitable for imaging and flow cytometry.
  • Amine-reactive NHS (succinimidyl) ester enables covalent labeling of primary amines.
  • Compatible with proteins, peptides, and amine-modified oligonucleotides.
  • High purity (95.0%) for reliable conjugation performance.
  • Supplied as a solid; store sealed at -20°C, protected from moisture and light.
  • COA, SDS, and handling documents available to support safe use and quality verification.