Learn More
Biolegend Alexa Fluor 700 anti-human/mouse/rat CD278 (ICOS)
Supplier:  Biolegend 313528
Encompass_Preferred
Alexa Fluor 700 anti-human/mouse/rat CD278 (ICOS) [C398.4A]; Isotype: Armenian Hamster IgG; Reactivity: Human, African Green, Baboon, Cynomolgus, Mouse, Rat, Rhesus, Swine (Pig, Porcine); Apps: FC; Size: 100 ug
Catalog No. NC1022561