Learn More
Biolegend Alexa Fluor 700 anti-mouse IFN-γ
Supplier:  Biolegend 505824
Encompass_Preferred
Alexa Fluor 700 anti-mouse IFN-γ [XMG1.2]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 100 ug
Catalog No. NC1253831