Learn More
Abcam Ale x a Fluor 700 Conjugation Kit (Fast) - Lightning-Link, 1MG
Catalog No. 502656323
Encompass_Preferred
Alexa Fluor 700 conjugation / labeling in < 20 mins with 30 secs hands-on time using Alexa Fluor 700 Conjugation Kit (Fast) - Lightning-Link ab269824. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-6323 Supplier Abcam Supplier No. AB2698241MG
Alexa Fluor 700 conjugation / labeling in < 20 mins with 30 secs hands-on time using Alexa Fluor 700 Conjugation Kit (Fast) - Lightning-Link ab269824. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement